Learn More
Thermo Scientific™ Accessories for Orion™ pH Electrodes
Product Code. 11757797
Change view
Click to view available options
For Use With (Equipment):
12 mm Electrodes, Glass
6 mm Electrode, Glass
8 mm Electrode, Glass
Sure-Flow Electrode, Standard Glass Electrode
Type:
Bulb Guard
Electrode Storage Bottle
Unit Size:
Pack of 3
Pack of 5
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Type Unit Size
11757797 Sure-Flow Electrode, Standard Glass Electrode Bulb Guard Pack of 5
10614131 12 mm Electrodes, Glass Electrode Storage Bottle Pack of 3
11727767 8 mm Electrode, Glass Electrode Storage Bottle Pack of 3
11737767 6 mm Electrode, Glass Electrode Storage Bottle Pack of 3
18897956 12 mm Electrodes, Glass Electrode Storage Bottle -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11757797 Supplier Thermo Scientific™ Supplier No. 910005
  • ONLINE EXCLUSIVE PRICE
    48.19 €
    39.42 € / Pack of 5
    Save 8.77 €
    18% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Store and protect your pH electrodes with these accessories.
Store and protect your pH electrodes with the Thermo Scientific™ Accessories for Orion™ pH Electrodes. Storage bottles protect and condition electrodes when not in use. Bulb guard protects the pH-sensing bulb of glass pH electrodes.
Electrode Bulb Guards
  • Designed for glass body pH electrodes
  • Use with standard or Thermo Scientific™ Orion™ Sure-Flow™ pH electrodes — one end of the bulb guard fits over standard electrodes while the other end fits over Sure-Flow electrodes without interfering with the Sure-Flow flushing action
TRUSTED_SUSTAINABILITY
Specifications
Description Bulb guard for glass pH Electrodes
For Use With (Equipment) Sure-Flow Electrode, Standard Glass Electrode
Type Bulb Guard