missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Accessories and Upgrade Kits for Varioskan LUX Multimode Microplate Reader

Product Code. 11995474 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
1536-well Adapter for 10 mm Plate w/o Lid
384-well Adapter for Plate w/ Lid
384-well Adapter for Plate w/o Lid
6-48-well Adapter for Plate w/ Lid
6-48-well Adapter for Plate w/o Lid
96-well Adapter for Plate w/ Lid
96-well Adapter for Plate w/o Lid
96-well adapter for PCR plate without lid
Aspirate tube assembly for a dispenser
Dispenser syringe, 1 mL
Dispenser syringe, 5 mL
Dispensing tube assembly for a dispenser
Emission Filter
Emission Filter Pick-up Tool
Liquid Protection Tray
Microplate package for Varioskan LUX IQ/OQ
Robotic plate tray adapter for plates without lid
Robotic plate tray for plates with and without lids
Storage Holder for Plate Adapters
Universal plate tray for plates without lid
Upgrade Kit for Luminometry and TRF
Upgrade kit for luminometry and AlphaScreen
Upgrade kit for luminometry, AlphaScreen and TRF
Upgrade kit for two dispensers
Unit Size:
Each
24 product options available for selection
Product selection table with 24 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type unitSize
11995474 6-48-well Adapter for Plate w/ Lid Each
15330787 Storage Holder for Plate Adapters Each
15340787 Emission Filter Pick-up Tool Each
12433458 Aspirate tube assembly for a dispenser Each
12439757 Emission Filter Each
11599479 Robotic plate tray adapter for plates without lid Each
12472238 384-well Adapter for Plate w/o Lid Each
12871886 384-well Adapter for Plate w/ Lid Each
12492238 96-well Adapter for Plate w/o Lid Each
11905484 96-well Adapter for Plate w/ Lid Each
11589309 6-48-well Adapter for Plate w/o Lid Each
12432248 Universal plate tray for plates without lid Each
12442248 Robotic plate tray for plates with and without lids Each
12452248 96-well adapter for PCR plate without lid Each
12472248 1536-well Adapter for 10 mm Plate w/o Lid Each
12492248 Dispenser syringe, 5 mL Each
12402258 Dispensing tube assembly for a dispenser Each
12412258 Dispenser syringe, 1 mL Each
17198238 Microplate package for Varioskan LUX IQ/OQ Each
17954312 Liquid Protection Tray Each
17972051 Upgrade kit for luminometry and AlphaScreen Each
17982081 Upgrade Kit for Luminometry and TRF Each
17932161 Upgrade kit for luminometry, AlphaScreen and TRF Each
17982051 Upgrade kit for two dispensers Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 24 options available.
24 options
This item is not returnable. View return policy
Product Code. 11995474 Supplier Thermo Scientific™ Supplier No. N02697
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Various accessories for use with the Varioskan™ LUX Multimode Microplate Reader.

Thermo Scientific™ Varioskan™ LUX multimode microplate reader is a versatile tool for busy labs equipped with a flexible range of measurement technologies including Absorbance, Fluorescence Intensity, Luminescence, AlphaScreen, and Time-Resolved Fluorescence. You can configure the instrument to your needs, and when your research focus changes, just upgrade the instrument with service installable upgrade kits. Various plate adapters and other accessories helps you to optimize your assays.

  • Varioskan LUX is a modular instrument, you can upgrade it with missing features later.
    • Installation of the upgrade kits must be carried out by trained and authorized service personnel.
    • For TRF and/or AlphaScreen technology upgrades, the instrument must be equipped with a luminescence module.
    • TRF and/or AlphaScreen technology upgrades include excitation filters but not emission filters. Please order emission filters separately.
    • Please contact your local representative to discuss about the instrument upgrade options.
  • Plate adapters are used for adapting plates that are of different heights.
    • Adapters adjust the plates to the optimum height for measuring and dispensing.
    • Choose the adapter based on the plate format in use and whether the lid is needed or not.
  • There are two types of plate trays: universal and robotic.
    • The universal plate tray is for basic use and it is compatible with all plate formats (6- to 1536-well plates).
    • A robotic plate tray is for automated use with robots and is compatible with 96- to 1536-well plate formats.
  • If the dispensers are in frequent use, it is recommended to replace the aspirate and dispensing tubings once a year.
TRUSTED_SUSTAINABILITY

Specifications

Product Type 6-48-well Adapter for Plate w/ Lid
For Use With (Equipment) Varioskan LUX Microplate Reader
Plate Type 48-Well Plate
Product Line Varioskan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.