missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Accessories and Upgrade Kits for Varioskan LUX Multimode Microplate Reader

Product Code. 15330787 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
1536-well Adapter for 10 mm Plate w/o Lid
384-well Adapter for Plate w/ Lid
384-well Adapter for Plate w/o Lid
6-48-well Adapter for Plate w/ Lid
6-48-well Adapter for Plate w/o Lid
96-well Adapter for Plate w/ Lid
96-well Adapter for Plate w/o Lid
96-well adapter for PCR plate without lid
Aspirate tube assembly for a dispenser
Dispenser syringe, 1 mL
Dispenser syringe, 5 mL
Dispensing tube assembly for a dispenser
Emission Filter
Emission Filter Pick-up Tool
Liquid Protection Tray
Microplate package for Varioskan LUX IQ/OQ
Robotic plate tray adapter for plates without lid
Robotic plate tray for plates with and without lids
Storage Holder for Plate Adapters
Universal plate tray for plates without lid
Upgrade Kit for Luminometry and TRF
Upgrade kit for luminometry and AlphaScreen
Upgrade kit for luminometry, AlphaScreen and TRF
Upgrade kit for two dispensers
Unit Size:
Each
24 product options available for selection
Product selection table with 24 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type unitSize
15330787 Storage Holder for Plate Adapters Each
15340787 Emission Filter Pick-up Tool Each
12433458 Aspirate tube assembly for a dispenser Each
12439757 Emission Filter Each
11599479 Robotic plate tray adapter for plates without lid Each
12472238 384-well Adapter for Plate w/o Lid Each
12871886 384-well Adapter for Plate w/ Lid Each
12492238 96-well Adapter for Plate w/o Lid Each
11905484 96-well Adapter for Plate w/ Lid Each
11589309 6-48-well Adapter for Plate w/o Lid Each
11995474 6-48-well Adapter for Plate w/ Lid Each
12432248 Universal plate tray for plates without lid Each
12442248 Robotic plate tray for plates with and without lids Each
12452248 96-well adapter for PCR plate without lid Each
12472248 1536-well Adapter for 10 mm Plate w/o Lid Each
12492248 Dispenser syringe, 5 mL Each
12402258 Dispensing tube assembly for a dispenser Each
12412258 Dispenser syringe, 1 mL Each
17198238 Microplate package for Varioskan LUX IQ/OQ Each
17954312 Liquid Protection Tray Each
17972051 Upgrade kit for luminometry and AlphaScreen Each
17982081 Upgrade Kit for Luminometry and TRF Each
17932161 Upgrade kit for luminometry, AlphaScreen and TRF Each
17982051 Upgrade kit for two dispensers Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 24 options available.
24 options
This item is not returnable. View return policy
Product Code. 15330787 Supplier Thermo Scientific™ Supplier No. N16443
Discount Lab Supplies
Use promo code 28170
View all promotions
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Various accessories for use with the Varioskan™ LUX Multimode Microplate Reader.

Thermo Scientific™ Varioskan™ LUX multimode microplate reader is a versatile tool for busy labs equipped with a flexible range of measurement technologies including Absorbance, Fluorescence Intensity, Luminescence, AlphaScreen, and Time-Resolved Fluorescence. You can configure the instrument to your needs, and when your research focus changes, just upgrade the instrument with service installable upgrade kits. Various plate adapters and other accessories helps you to optimize your assays.

Varioskan LUX is paired with the powerful fourth generation SkanIt Software, providing excellent usability and flexibility.

  • License free—Each user can install the software on his/her computer
  • Intuitive and flexible interface for multiple users and assays
  • Innovative Virtual Pipetting ToolTM makes it easy to define samples to plate layout
  • Measurement data is continuously saved to the database, so there’s no data lost due to unexpected interruptions such as power outages or accidental aborting
  • Simplified data processing and export
  • Export data in a single mouse click, or have it emailed automatically at the end of the run
  • Several file formats for exporting results: *.xlsx, *.pdf, *.xml, and *.txt

The Varioskan LUX is modular and upgradeable, so you can build the system that best suits your needs. For more information, contact a sales representative.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Storage Holder for Plate Adapters
For Use With (Equipment) Varioskan LUX Microplate Reader
Product Line Varioskan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.