Learn More
Agilent BioTek Accessory for EL406 Microplate Washer and MultiFlo FX Microplate Dispenser
Shop All Agilent BioTek Products
Click to view available options
For Use With (Equipment):
2 Syringe Pump Dispensers, EL406 Microplate Washer, Agilent BioTek MultiFlo FX Multimode Dispenser
Unit Size:
Each
Description
- Spare syringe pump dispense tubing and check valve kit
- Organic solvent compatible (non-autoclavable, but compatible with the autoclavable syringe module hardware)
Specifications
Specifications
| For Use With (Equipment) | 2 Syringe Pump Dispensers, EL406 Microplate Washer, Agilent BioTek MultiFlo FX Multimode Dispenser |