Learn More
Cytiva Accessory Kits
Product Code. 11643224 Shop All Cytiva Products
Click to view available options
For Use With:
For HiScale 26 Columns
For HiScale 50 Columns
Unit Size:
Each
Product Code. 11643224
Brand: Cytiva 28966375
Only null left
Add to basket
Add to basket
Specifications
Type Accessory Kits
Packaging Quantity 1 Each
Includes Net ring, support screen, o-ring, two ferrules and two finger tight stop plugs
For Use With For HiScale 50 Columns