missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Acclaim™ 300 C18 HPLC Columns

Use these wide-pore C18 columns to achieve high-resolution protein separations.

Brand:  Thermo Scientific™ 069697

193.89 EUR valid until 2024-05-31
Use promo code "21664" to get your promotional price.



Code : 16

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 11321923

  • 204.09 € / Pack of 2
Estimated Shipment: 25-07-2024
to see stock.
Add to basket

Description

Description

Achieve high-resolution peptide mapping and protein separations with Thermo Scientific™ Acclaim™ 300 C18 LC Columns. The 3μm ultrapure silica particles reduce the limits for diffusion of the analytes into the stationary phase, resulting in fast, high-resolution separations. Compared to 5μm column packings, the smaller particles support increased flow rates and shallower gradients on shorter columns for faster separations.

High-Resolution Separation of Proteins and Peptides
  • High-performance C18 bonding chemistry on 300Å pore silica
  • Application tested for suitability in peptide mapping
  • Reproducible for dependable results
  • Extremely low bleed, fully compatible with MS
  • Stable between pH 2 .5 and 7.5
About the Acclaim Family of HPLC Columns
  • Complementary selectivity, high column efficiencies and ultrahigh resolution
  • Broad spectrum of stationary phases, including conventional C8 and C18 phases, polar embedded phases and specialty phases including wide-pore
  • Performance tested to ensure high quality and reliability
Specifications

Specifications

4.6 mm
LC/MS
6000 psi (414 bar)
2.5 to 7.5
100 m²/g
L1
Acclaim
8%
Yes
Guard Cartridge
1 mL/min
10 mm
3 μm
300 Å
60°C
C18
Ultrapure Silica
Reversed Phase
Reversed Phase
Bio-LC
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.