missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACCN5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
375.00 € - 515.00 €
Specifications
| Antigen | ACCN5 |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
ACCN5 Polyclonal specifically detects ACCN5 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| ACCN5 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 51802 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:EVQYVEKMEFPAVTFCNLNRFQTDAVAKFGVIFFLWHIVSKVLHLQEITANSTGSREATDFAASHQNFSIVEFIRNKGFYLNNSTLLDCEFFGKPCSPKDFAHVFTEYGNCFTFNHGETLQAKRKVSVSGRGLSLLFNVNQEAFTDNPAL | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| amiloride-sensitive cation channel 5, intestinal | |
| ASIC5 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title