missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Accu20 Ultrapure Water System

Product Code. 15489689 Shop All Fisherbrand Products
Change view
Click to view available options
Product Type:
ACCU20 UV Ultrapure Water System
ACCU20 Ultrapure Water System
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type unitSize
15489689 ACCU20 Ultrapure Water System Each
15499689 ACCU20 UV Ultrapure Water System Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15489689

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Fisherbrand™ Accu20 Ultrapure Water System used to produce Type I ultrapure water and RO water on demand from tap water directly.

  • Compact system provides easy way to produce ultrapure water or RO water on demand from tap water directly with no additional pre-treatment filters
  • Equipped with safeguard AccuDuo P pack cartridge and dual-column AccuDuo U Pack ultrapurification cartridge to ensure optimum quality of final product water
  • No external storage tank required, which eliminate risk of water degration by contaminants from container or tubing
  • Fully automatic controlled and maintained system contains automatic cleaning cycles and selfmaintenance functions for RO membrane, which increases lifetime and efficiency of RO membranes as well as cartridges
  • Automatic hourly system recirculation in loop prevents bacterial growth and maintain water purity at all times
  • System status and key quality parameters are clearly displayed on large LCD screen for easy system performance monitoring

Applications:

  • HPLC mobile phase preparation
  • Preparation of reagent blank solutions
  • As sample diluent for GC, HPLC, AA, ICP-MS and other analytical techniques
  • Preparation of buffers and culture media for mammalian cell culture
  • Preparation of molecular biology reagents, etc.

Certifications

Meets or exceeds ASTM, CLSI, CAP, and ISO Type I water standards

Specifications

Conductivity RO water at 25°C: Typically <20μS/cm
Bacterial Content <0.1 CFU/mL (with a 0.2 μm final filter or terminal ultrafiltration cartridge )
Flow Rate RO water (at 25°C ): >0.5L/min.; Ultrapure water ( Type I ) (at 25°C ): >0.5L/min.
Resistivity 18.2 Mohm.cm
TOC Monitor with UV Intensity Monitoring <10 ppb (<5 ppb with a dual wavelength UV lamp)
Feedwater Pressure 15 to 90 psi (1 to 6 bar )
Model Accu20
Particle Size <1/mL (with a 0.2μm final filter or terminal ultrafiltration cartridge)
Temperature Range (Metric) 5°C to 45°C
Length (English) 76.2 in.
Length (Metric) 30 cm
Height (English) 18.8 in.
Height (Metric) 48 cm
Depth (English) 20 in.
Depth (Metric) 51 cm
Product Type ACCU20 Ultrapure Water System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.