missing translation for 'onlineSavingsMsg'
Learn More

Fisher Scientific™ Accu500 Water System

Product Code. 15479699 Shop All Fisherbrand Products
Change view
Click to view available options
Product Type:
UV Ultrapure Water System
UV Ultrapure Water System w/ Dispenser
Ultrapure Water Purification System
RO Flow Rate:
10 L/hr.
15 L/hr.
15 L/min.
20 L/hr.
30 L/hr.
Unit Size:
Each
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type RO Flow Rate unitSize
15479699 UV Ultrapure Water System 30 L/hr. Each
15409699 Ultrapure Water Purification System 10 L/hr. Each
15419699 UV Ultrapure Water System 10 L/hr. Each
15409709 UV Ultrapure Water System w/ Dispenser 10 L/hr. Each
15429699 Ultrapure Water Purification System 15 L/hr. Each
15439699 UV Ultrapure Water System 15 L/min. Each
15419709 UV Ultrapure Water System w/ Dispenser 15 L/hr. Each
15449699 Ultrapure Water Purification System 20 L/hr. Each
15459699 UV Ultrapure Water System 20 L/hr. Each
15429709 UV Ultrapure Water System w/ Dispenser 20 L/hr. Each
15469699 Ultrapure Water Purification System 30 L/hr. Each
15439709 UV Ultrapure Water System w/ Dispenser 30 L/hr. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Product Code. 15479699

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Exceeds ASTM, CLSI, CAP, and ISO Type I water standards. Fisher Scientific™ Accu500 Water System is a versatile purification system that can be used in a number of ways.

  • The fully automated control system contains pre-rinse, a flush feature preventing RO membrane fouling, RO membrane and cartridge life monitoring as well as many other programs for convenient operation
  • The pre-rinsing program starts automatically once a new RO membrane is installed so there is no need for user to set additional cleaning procedures
  • Two sets of dual-column purification cartridges work sequentially to ensure thorough removal of ions and organics in water
  • Automatic hourly system recirculation prevents bacterial growth
  • Resistivity is measured by a highly sensitive resistivity sensor
  • Electrical and water compartments are completely segregated, thus safer to operate
  • User can set water dispensing time for easy water collection
  • Both RO and ultrapure water can be dispensed from the water system
  • The system can be fitted with a 30L or 60L PE tank with a liquid level sensor and a tank vent filter with CO2 scavenger to protect purified water from atmospheric degradation
  • A 0.2μm filter or ultrafiltration filter (optional) removes final contaminants
  • Independent ultrapure water dispenser
  • The remote dispenser has a built-in high precision resistivity sensor with cell constant of 0.01cm-1 to measure the quality of ultrapure water just prior to the dispensing point
  • Volumetric dispensing and calibration
  • The handle is 360 degree-rotatable and detachable from its support for effortless and instant dispensing with maximum flexibility

Applications:
Ultrapure and RO water can be used in many areas. Some typical applications include:

Ultrapure water

  • HPLC mobile phase preparation
  • Preparation of reagent blank solutions
  • As sample diluent for GC, HPLC, AA, ICP-MS and other analytical techniques
  • Preparation of buffers and culture media for mammalian cell culture
  • Preparation of molecular biology reagents, etc.

RO water

  • Glassware cleaning
  • Washing machine for glassware
  • Waterbath water
  • Autoclaving

Specifications

Conductivity <20μS/cm
Product Water Type Tap Water
Resistivity 18.2 Mohm.cm
Feedwater Pressure 1 to 6 bar
Model Accu500
Particles >0.2µM/mL <1/mL with 0.2μm final filter or terminal ultrafiltration cartridge
Purified Water Flowrate Field 1.5 L/min.
RO Flow Rate 30 L/hr.
Temperature Range (Metric) 5°C to 45°C
Length (Metric) 30 cm
Height (Metric) 48 cm
Depth (Metric) 51 cm
Endotoxin Concentration <0.001 EU/mL with a terminal ultrafiltration cartridge
Product Type UV Ultrapure Water System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.