Learn More
Beschreibung
Beinhaltet
accumet AB150 pH benchtop meter, TRIS-compatible double junction refillable glass-body pH electrode (13-620-183A), ATC temperature probe (13-620-19), electrode stand, RS232 and USB cables, 100/240V universal power supply
Provide intuitive, simple operation and high accuracy measurements in a compact, affordable meter. Fisherbrand accumet AB150 pH benchtop meters have large backlit display which offers great visibility in laboratory environments. The accumet AB150 pH meter is an industry favorite for when reliability and consistency in measurements matter.
- Graphic backlit display with date and time, stability indicator and alarms
- One to six point pH calibration with 0.001 pH resolution
- Automatic buffer recognition with four pH buffer sets or custom buffer option
- pH slope and offset display
- One point relative mV calibration
- One point ATC temperature probe calibration
- Automatic or manual temperature compensation, displayed in °C or °F
- Password protection of calibration and setup modes
- 500 data set storage
- RS232 and USB interfaces
- Multilanguage interface with English, German, Chinese, French, Italian and Spanish
- Optional stirring probe (Cat. No. 13-620-BSP) with five speed settings
NIST-Traceable Calibration Models (-CERT)
- Help meet ISO, FDA, USDA and EPA quality assurance guidelines
- Includes meter accuracy at five test points across pH range
- Includes meter accuracy at five test points across corresponding mV range
- Testing performed by accredited laboratory; includes NIST-Traceable certificate
Specifications
- pH Range: -2.000 to 20.000 pH
- pH Resolution: 0.1, 0.01, 0.001 pH
- pH Accuracy: ±0.002 pH
- mV Range: ±2000.0mV
- mV Resolution: 0.1mV
- mV Accuracy: ±0.2mV or ±0.05 %, whichever is greater
- mV Offset Adjustment: Up to ±150mV
- Temperature Range: 0° to 100°C (32° to 212.0°F)
- Temperature Resolution: 0.1°C (0.1°F)
- Temperature Accuracy: ±0.3°C (±0.5°F)
- Calibration Points: 1 to 6 points with automatic buffer recognition (ABR)
- ATC Temperature Calibration: Offset in 0.1°C increments; Offset range: ±5°C (±9°F)
- Self-Test: Yes
- Inputs: BNC, mini-phone ATC, ground
- Power Requirements: 100 to 240VAC, 50/60Hz
Applications
Academic, research, QA/QC, drinking water, wastewater, food, beverage, industrial
Zertifizierungen
Select models available with Certificate of NIST Traceability
Garantie
3 years on meter
Spezifikation
Spezifikation
| Includes | Meter, TRIS Compatible accuTupH pH Electrode (13-620-183A), ATC Probe (13-620-19), Electrode Arm, 110/220V Power Supply |
| Accuracy (Temperature) | ±0.3°C |
| Accuracy (mV) | ±0.2 |
| Accuracy (pH) | ±0.002 |
| Data Management Details | Manual, Timed (selectable every 3 to 3600 sec.), Printer or CSV Format |
| Outputs | RS-232 (phono plug), mini-B USB, stirrer |
| Range (pH) | -2 to 20 |
| Resolution (pH) | 0.001, 0.01, 0.1 |
| Calibration Points (pH) | Up to 6 pre-set or 5 custom |
| Range (mV) | ±2000 |
| Mehr anzeigen |
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.