missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ accumet™ AB250 Benchtop pH/ISE Meters

Product Code. 12850633 Shop All Fisherbrand Products
Click to view available options
Product Type:
AB250 pH/ISE Meter
Meter with pH/ISE Kit
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12850633

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

accumet AB250 pH/ISE benchtop meter, electrode stand, RS232 and USB cables, 100/240V universal power supply

Provides intuitive, simple operation and high accuracy measurements in a compact, affordable meter. Fisherbrand accumet AB250 pH/ISE meter provides flexible measurements of pH or ion concentration using an ion selective electrode (ISE) in one easy to use, reliable measurement system.

Provides intuitive, simple operation and high accuracy measurements in a compact, affordable meter. Fisherbrand accumet AB250 pH/ISE meter provides flexible measurements of pH or ion concentration using an ion selective electrode (ISE) in one easy to use,

  • Graphic backlit display with date and time, stability indicator and alarms
  • One to six point pH calibration with 0.001 pH resolution
  • Automatic buffer recognition with four pH buffer sets or custom buffer option
  • pH slope and offset display
  • Two to six point ISE calibration with 0.001 ppm, mg/L or molar resolution
  • Automatic or manual temperature compensation, displayed in °C or °F
  • Password protection of calibration and setup modes
  • 500 data set storage
  • RS232 and USB interfaces
  • Multilanguage interface with English, German, Chinese, French, Italian and Spanish
  • Optional stirring probe (Cat. No. 13-620-BSP) with five speed settings

NIST-Traceable Calibration Model (-CERT)

  • Help meet ISO, FDA, USDA and EPA quality assurance guidelines
  • Includes meter accuracy at five test points across pH range
  • Includes meter accuracy at five test points across corresponding mV range
  • Testing performed by accredited laboratory, includes NIST-Traceable certificate

Specifications

  • pH Range: -2.000 to 20.000 pH
  • pH Resolution: 0.1, 0.01, 0.001 pH
  • pH Accuracy: ±0.002 pH
  • ISE Range: 0.001 to 19999 ppm, mg/L or molar
  • ISE Resolution: 0.001, 0.01, 0.1, 1 (automatic)
  • ISE Accuracy: 0.5 % full scale (monovalent ion), 1 % full scale (divalent ion)
  • mV Range: ±2000.0mV
  • mV Resolution: 0.1mV
  • mV Accuracy: ±0.2mV or ±0.05 %, whichever is greater
  • mV Offset Adjustment: Up to ±150mV
  • Temperature Range: 0° to 100°C (32° to 212.0°F)
  • Temperature Resolution: 0.1°C (0.1°F)
  • Temperature Accuracy: ±0.3°C (±0.5°F)
  • Calibration Points: 1 to 6 points with automatic buffer recognition (ABR)
  • ATC Temperature Calibration: Offset in 0.1°C increments; Offset range: ±5°C (±9°F)
  • Self-Test: Yes
  • Inputs: BNC, mini-phone ATC, ground
  • Power Requirements: 100 to 240VAC, 50/60Hz

Applications
Academic, research, QA/QC, drinking water, wastewater, food, beverage, industrial

Certifications

Select models available with Certificate of NIST Traceability

Warranty

3 years on meter

TRUSTED_SUSTAINABILITY

Specifications

Outputs RS232 (phono plug), mini-B USB, stirrer
Accuracy (ISE) 0.5% full scale (monovalent ion), 1% full scale (divalent ion)
Accuracy (mV) ±0.2 mV
Accuracy (pH) ±0.002
Calibration Points (ISE) 2 to 6 points from one of following group, 0.001, 0.01, 0.1, 1, 10, 100, 0.01, 0.1, 1, 10, 100, 1000
Calibration Points (pH) Up to 6 preset or 5 custom
Certifications/Compliance Without NIST-traceable pH Certificate
Data Management Details Manual, timed (selectable every 3 to 3600 sec.), printer or CSV format
Data Points 500
Display Type LCD
Measurement Modes pH, mV, ISE, Ion, Concentration, Temperature
Range (mV) ±2000 mV
Range (pH) -2 to 20
Resolution (ISE) 0.001/0.01/0.1/1 (automatic)
Resolution (pH) 0.1/0.01/0.001
Temperature Range (Metric) 0°C to 100°C
Includes Meter, Electrode Arm, RS-232 and USB Cables, 100/240V Power Supply
Inputs BNC, ATC
Model AB250 pH/ISE Meter Only
Product Type AB250 pH/ISE Meter
Range (ISE) 0.001 to 19999 (±2000)
Resolution (mV) 0.1mV
Temperature Accuracy ±0.3°C
Temperature Resolution 0.1°C
Warranty 3 Years
Voltage 100/240 V
Frequency 50/60Hz
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.