Learn More
Fisherbrand™ accumet™ AB315 and AB330 Meter Replacement Universal Power Adapter
Product Code. 17358949 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
For Use With (Equipment):
accumet AB315 and AB330 Meters
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
17358949 accumet AB315 and AB330 Meters Each
1 options
Product Code. 17358949
  • ONLINE EXCLUSIVE PRICE
    52.60 €
    47.41 € / Pack of 1
    Save 5.19 €
    9% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Select accessories for your Fisherbrand™ accumet™ AB315 pH/mV meter and Fisherbrand™ accumet™ AB330 conductivity meter.
  • Universal 100/240 V 50/60 Hz power adapter
  • Includes wall plugs for US/Japan, Euro, UK/Singapore, Australia/New Zealand and China
TRUSTED_SUSTAINABILITY
Specifications
Type Replacement universal power adapter
Frequency 50/60 Hz
Includes Wall plugs for US/Japan, Euro, UK/Singapore, Australia/New Zealand and China
Voltage 100/240 V
Compatibility accumet™ AB300 series meters
For Use With (Equipment) accumet AB315 and AB330 Meters