Learn More
Description
Designed for routine pH measurements in the lab with convenience in mind – simple functionality, stored data sets in a non-volatile memory and meter-attachable electrode stand. The accumet AE150 pH meters offer efficient sample reading and recording with flexible display options to show display both pH and temperature or mV and temperature simultaneously.
- One to five point pH calibration
- Includes 13-620-AE6 3-in-1 single junction gel pH/ATC electrode
- Automatic buffer recognition with two pH buffer sets
- pH slope and offset display
- One point mV calibration
- One point ATC temperature probe calibration
- Automatic or manual temperature compensation
- Ready icon to indicate measurement stability
- Electrode condition icon to indicate status
- 100 data set storage with non-volatile memory
Warranty
3 years on meter
Specifications
Specifications
| Includes | Meter, 13-620-AE6 3-in-1 single-junction gel pH/ATC electrode, and stand |
| Accuracy (Temperature) | ±0.3°C |
| Accuracy (mV) | ±0.2 ±199.9 ±2 beyond ±200 |
| Accuracy (pH) | ±0.01 |
| Outputs | Recorder (±2000 mV) |
| Range (pH) | 0 to 14 |
| Resolution (pH) | 0.01 |
| Range (mV) | ±1999 |
| Resolution (mV) | 0.1 mV (±199.9 mV); 1 mV beyond ±200 mV |
| Calibration Points | 1 to 5 with automatic buffer recognition (ABR) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.