missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ accumet™ AR415 pH Bench Meter

Product Code. 31702804 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Model:
AR415
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Unit Size
31702804 AR415 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31702804
  • ONLINE EXCLUSIVE PRICE
    1560.00 €
    773.01 € / 1 sets
    Save 786.99 €
    50% Off
View all promotions
Estimated Shipment: 17-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed to deliver precise and reliable electrochemistry testing, the accumet AR415 offers enhanced performance in a streamlined design. Ideal for laboratories requiring dependable measurement capabilities.

  • Large 7-in. high-resolution color display with capacitive touchscreen operable with gloves.
  • One to five point pH calibration with automatic buffer recognition.
  • Streamlined, easy-to-operate user interface with step-by-step calibration guidance.
  • Stores 10 calibration logs per parameter with date and time.
  • Stores 5000 data log sets with date and time.
  • Outputs to a printer, computer, or USB flash drive for long-term data retention.
  • Inputs sample ID with optional USB barcode scanner, USB keyboard, or USB mouse.
  • Stirs samples with meter-controlled and meter-powered stirrer probe.
TRUSTED_SUSTAINABILITY

Specifications

Includes pH Meter
Accuracy (Temperature) ±0.3°C
Accuracy (mV) -2000.0 to 2000.0 mV
Accuracy (pH) ±0.002 pH
Measurement Modes pH, ORP, mV, Temperature
Calibration pH, Relative mV, Temperature
Range (pH) -2 to 20
Resolution (pH) 0.1, 0.01, or 0.001 pH
Range (mV) -2000.0 to 2000.0 mV
Calibration Points 1
Temperature Range (Metric) -30.0 to +130.0°C
Display Type 7-in. High-resolution Color Graphic LCD
Backlight Option Yes
Data Points 5000 with Date and Time Stamp
Log Function Type Automatic Data Logging
Certifications/Compliance CE, TUV 3-1, FCC Class A
IP Rating IP54
Model AR415
Channels 1
Type pH Meter
Voltage 100/240 V
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.