missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ accumet™ AR425 pH/Ion Meter pH Kit

Product Code. 31702818 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Model:
AR425A
AR425B
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Unit Size
31702818 AR425A 1 set
31702819 AR425B 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 31702818
  • ONLINE EXCLUSIVE PRICE
    1741.67 €
    1180.73 € / 1 sets
    Save 560.94 €
    32% Off
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed to deliver precise and reliable electrochemistry testing, the accumet AR425 offers enhanced performance in a streamlined design. Ideal for laboratories requiring dependable measurement capabilities.

  • Large 7-in., high-resolution color display with capacitive touchscreen operable with gloves.
  • One to five point pH and Ion calibration with automatic buffer/standard recognition.
  • Streamlined, easy-to-operate user interface with step-by-step calibration guidance.
  • Stores 10 calibration logs per parameter with date and time.
  • Stores 5000 data log sets with date and time.
  • Outputs to a printer, computer, or USB flash drive for long-term data retention.
  • Inputs sample ID with optional USB barcode scanner, USB keyboard, or USB mouse.
  • Stirs samples with meter-controlled and meter-powered stirrer probe.
TRUSTED_SUSTAINABILITY

Specifications

Accuracy (mV) ±0.2 mV or ±0.05% of the reading, whichever is greater
Accuracy (pH) ±0.002 pH
Backlight Option Yes
Calibration pH
Calibration Points 1
Certifications/Compliance CE, TUV 3-1, FCC Class A
Data Points 5000 with Date and Time Stamp
Channels 1
Display Type 7-in. High-resolution Color Graphic LCD
IP Rating IP54
Measurement Modes pH, mV, ORP, Ion, Temperature
Range (mV) -2000.0 to +2000.0 mV
Range (pH) -2 to 20
Resolution (ISE) 0.0001 to 19999
Resolution (pH) 0.1, 0.01, or 0.001 pH
Temperature Range (Metric) -30.0 to 130.0°C
Log Function Type Automatic Data Logging
Model AR425A
Product Type Benchtop Multiparameter Meters
Voltage 100/240 V
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.