missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ accumet™ AR450 pH/ion/Conductivity Meter Kit

Product Code. 31702817 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Product Type:
pH/Ion/Conductivity Meter
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
31702817 pH/Ion/Conductivity Meter 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31702817
  • ONLINE EXCLUSIVE PRICE
    3545.00 €
    1758.07 € / 1 sets
    Save 1786.93 €
    50% Off
View all promotions
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed to deliver precise and reliable electrochemistry testing, the accumet AR450 offers enhanced performance in a streamlined design. Ideal for laboratories requiring dependable measurement capabilities.

  • Large 7-in. high-resolution color display with capacitive touchscreen operable with gloves
  • One to five point pH, Ion, and conductivity calibration with automatic buffer/standard recognition
  • Streamlined, easy-to-operate user interface with step-by-step calibration guidance
  • Stores 10 calibration logs per parameter with date and time
  • Stores 5,000 data log sets with date and time
  • Outputs to a printer, computer, or USB flash drive for long-term data retention
  • Inputs sample ID with optional USB barcode scanner, USB keyboard, or USB mouse
  • Stirs samples with meter-controlled and meter-powered stirrer probe
TRUSTED_SUSTAINABILITY

Specifications

Accuracy (Conductivity) ±1% reading >3 μS/cm; ±1% reading ±0.01 μS/cm ≤3 μS/cm
Accuracy (Ion) ±0.2 mV or ±0.05% of the reading, whichever is greater
Accuracy (mV) ±0.2 mV or ±0.05% of the reading, whichever is greater
Accuracy (pH) ±0.002 pH
Accuracy (Resistivity) ±1% reading
Accuracy (Salinity) ±1% reading
Accuracy (TDS) ±1% reading
Calibration Points 2
Certifications/Compliance CE, TUV 3-1, FCC Class A
Data Points 500 with Date and Time Stamp
Channels 2
Display Type 7-in. High-resolution Color Graphic LCD
IP Rating IP54
Measurement Modes pH, mV, Ion, Concentration, TDS, Salinity, Resistivity, Temperature
Range (Conductivity) 0.010 μS/cm to 3000 mS/cm
Range (mV) -2000.0 to 2000.0 mV
Range (pH) -2 to 20
Range (Resistivity) 1.0 Ω to 100.0 MΩ
Range (Salinity) 1.0 Ω to 100.0 MΩ
Range (TDS) 0.010 ppm to 500.0 ppt (TDS factor of 1.00)
Resolution (ISE) 0.0001 to 19999
Resolution (pH) ±0.002 pH
Temperature Range (Metric) -30.0 to 130.0°C
Product Type pH/Ion/Conductivity Meter
Range (Ion) 0.0001 to 19999
Resolution (Conductivity) 0.001 μS/cm minimum, auto-ranging, up to 4 digits
Resolution (Ion) 0.0001 minimum, 1 to 5 digits
Resolution (mV) 0.1 mV
Resolution (Resistivity) 0.1 Ω minimum, auto-ranging
Resolution (Salinity) 0.001 ppt minimum, auto-ranging
Resolution (TDS) 0.001 ppm minimum, auto-ranging, up to 4 digits
Temperature Accuracy ±0.3°C, ±0.5°F
Temperature Resolution 0.1°C, 0.1°F
Voltage 100/240 V
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.