missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ accumet™ Glass Body Standard Size Combination Electrodes - Mercury-Free

Product Code. 10232781 Shop All Fisherbrand Products
Click to view available options
Connector Types:
BNC
U.S. Standard/Pin
Junction:
Double Ceramic
Single Ceramic
Filling Solution Type:
SP135-500
SP138-500
Length (Metric) Cable:
76.2 cm
152.4 cm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10232781

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Includes

Removable bulb protector

For routine pH and titration measurements with small samples and small vessels, where separate half-cell electrodes may be impractical

 

  • 0 to 14pH range
  • Porous ceramic junction
  • Twist-open/twist-close cap for easy filling
  • Constant distance between FS-5 Universal Glass pH bulb and reference junction means high reproducibility
  • Immersion depth, ∽4 in. (10.1cm)
  • 30 in. long (76.2cm) leads, except 13-620-91, which has 60 in. long (152.4cm) lead

 

Choice of Ag/AgCl Single- or Double-Junction internal reference elements

  • Single-junction models with Ag/AgCl reference are perfect for routine pH and titration measurements of aqueous solutions with small samples, small vessels
  • Double-junction models with Ag/AgCl reference are useful in aqueous and nonaqueous solutions, and in applications requiring TRIS buffers
  • For aqueous and nonaqueous samples
  • Double-junction Ag/AgCl eliminate measurement and clogging problems when used in research with Tris buffers

 

TRUSTED_SUSTAINABILITY

Specifications

Closure Type Twist Open/Twist Close
Color Blue
Connector Types BNC
Junction Single Ceramic
Material Glass
pH Range 0 to 14
Temperature (Metric) Operating 23°C to 212°C
Type Combination pH Electrode
Filling Solution Type SP135-500
For Use With (Application) For routine pH and titration measurements with small samples and small vessels, where separate half cell electrodes may be impractical
For Use With (Equipment) pH meter
Includes Removable bulb protector
Internal Reference Ag/AgCl
Response Time <10 sec.
Temperature (English) Operating -5°F to +100°F
Temperature (Metric) Range -5°C to +100°C
Warranty 1 Year
Length (English) 4 in.
Length (English) Cable 30 in.
Length (Metric) 101.6 mm
Length (Metric) Cable 76.2 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.