missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ AccuSEQ™ Real-Time PCR Detection Software v2.2

Product Code. 18753634 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18753634 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18753634 Supplier Applied Biosystems™ Supplier No. A58642

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

AccuSEQ Real-Time PCR Detection Software v2.2 is part of an integrated workflow for impurity and contaminant testing during biopharmaceutical manufacturing using the SEQ family of real-time PCR assays, including MycoSEQ, resDNASEQ and ViralSEQ assays.

AccuSEQ Real-Time PCR Detection Software v2.2 is part of an integrated workflow for impurity and contaminant testing during biopharmaceutical manufacturing using the SEQ family of real-time PCR assays, including MycoSEQ, resDNASEQ and ViralSEQ assays. The software is designed to work with the Applied Biosystems 7500 Fast Real-Time PCR Instrument with 0.1-mL, 96-well plates, and Windows 10 operating system. This software is provided via a link for online download.

Features of AccuSEQ Real-Time PCR Detection Software include:
• Accurate host cell impurities quantitation for resDNASEQ Host Cell Residual DNA Quantitation assays
• Automated presence/absence calls for MycoSEQ Mycoplasma and ViralSEQ MMV/Vesivirus assays
• Accurate quantitation of Sf-rhabdovirus genomic RNA for ViralSEQ Quantitative Sf-Rhabdovirus Kit
• Single software platform for all current and future SEQ real-time PCR assays
• Security, audit, and e-signature capabilities to help enable 21 CFR Pt 11 compliance

Residual DNA quantitation
For host cell residual DNA quantitation, a custom module with next-generation algorithms can be used with the resDNASEQ CHO Residual DNA Assay for accurate quantitation.

Mycoplasma detection
For rapid and accurate Mycoplasma detection, AccuSEQ software offers a plug-in analysis module for automated presence/absence calls when using the MycoSEQ Mycoplasma Detection system. Automated analysis tools enable one-click processing of MycoSEQ assay data, helping deliver presence or absence calls within seconds of data collection being completed.

MMV and vesivirus detection
AccuSEQ software is optimized and validated for the ViralSEQ MMV and Vesivirus Detection assay. The software’s visual result summary provides immediate understanding of presence or absence of virus in your samples.

Sf-rhabdovirus genomic RNA quantitation
AccuSEQ software helps enable rapid and easy analysis of ViralSEQ Quantitative Sf-Rhabdovirus system data without the hassle of multiple exporting steps and reliance on third party software.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) 7500 Fast System
Product Line AccuSEQ
Software Type Real-Time PCR Software
Product Type Real Time PCR Detection Software
For Use With (Application) Real Time PCR (qPCR)
Shipping Condition Room Temperature
Quantity Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.