missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ AccuSEQ™ Real-Time PCR Detection Software v3.1
Shop All Applied Biosystems ProductsSpecifications
Specifications
| Content And Storage | Product letter and License registration code kit |
| For Use With (Equipment) | QuantStudio™ 5, 0.1 mL Block Instrument |
| Product Line | AccuSEQ |
| Software Type | Real-Time PCR Software |
| Product Type | Real Time PCR Detection Software |
| For Use With (Application) | Real Time PCR (qPCR) |
| Shipping Condition | Room Temperature |
| Quantity | 1 Kit |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?