missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Accutase™ Cell Detachment Solution, Liquid
BIOPHARMA

Product Code. 15323609
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15323609 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15323609 Supplier Corning™ Supplier No. 25058CI
  • ONLINE EXCLUSIVE PRICE
    87.54 €
    49.16 € / 100mL
    Save 38.38 €
    43% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Efficient, with gentle detachment of adherent cells within minutes

Accutase is useful for the routine detachment of cells from standard tissue culture plasticware and adhesion coated plasticware, including our selection of Advanced Surface coated plasticware. Accutase is useful for cell detachment and for preparing single-cell suspensions from clumped cell populations for sub-culturing cells, analytical studies, and for accurate cell counting.

Accutase is formulated at a concentration that is ready to use, once thawed. (Note: Never thaw Accutase at 37°C.) A thawed bottle of Accutase can be removed from the refrigerator and immediately applied to cells. It should not be pre-warmed to 37°C. Accutase contains proteolytic activity that gently breaks down cellular adhesion molecules and enables cell detachment from the bottom of the flask.

Accutase cell detachment solution is:

  • Efficient, with gentle detachment of adherent cells within minutes
  • An alternative to Trypsin
  • Able to eliminate neutralization or wash steps
  • Accutase does not contain mammalian or bacterial derived products
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -20°C.
Form Liquid
Sterility Sterile
Formulation 1 X Accutase enzymes prepared in Dulbeccos PBS (0.2g/L KCl, 0.2g/L KH2PO5, 8g/L NaCl, and 1.15g/L Na2HPO4) containing 0.5 mm EDTA·4Na and 3 mg/L Phenol Red.
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.