Learn More
ACE-2 Goat anti-Mouse, Polyclonal, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody has been used in 6 publications
Brand:  R&D Systems AF3437-SP
Encompass
Product Code. 15792024
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
ACE-2 Polyclonal specifically detects ACE-2 in Mouse samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation.