missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ACEA

Product Code. p-200078696
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 18762774

Brand: Tocris Bioscience™ 1319/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and highly selective CB1 agonist

ACEA is a potent and highly selective CB1 receptor agonist (Ki = 1.4 nM). Displays >1400-fold selectivity over CB2 receptors. Active in vivo.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(2-Chloroethyl)-5Z,8Z,11Z,14Z-eicosatetraenamide
CAS 220556-69-4
Quantity 25 mg
Target Cannabinoid R1/CB1 Agonists
Molecular Formula C22H36ClNO
Purity 0.97
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.