missing translation for 'onlineSavingsMsg'
Learn More

Acemetacin, MedChemExpress

Product Code. 16490948
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16490948 5 g 5g
16422199 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16490948 Supplier Medchem Express Supplier No. HYB0482
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Acemetacin (TVX 1322) is a non-steroidal anti-inflammatory drug and a glycolic acid ester of indometacin that is a cyclooxygenase inhibitor.

Acemetacin (TVX 1322) is a non-steroidal anti-inflammatory drug and a glycolic acid ester of indometacin that is a cyclooxygenase inhibitor.

Specifications

Chemical Name or Material Acemetacin
CAS 53164-05-9
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C21H18ClNO6
Quantity 5 g
Synonym TVX 1322
Solubility Information DMSO : 125 mg/mL (300.61 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(OCC(O)=O)CC1=C(C)N(C(C2=CC=C(Cl)C=C2)=O)C3=C1C=C(OC)C=C3
Molecular Weight (g/mol) 415.82
Formula Weight 415.82
Percent Purity 99.95%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.