missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acenaphthylene-d8, CDN
Click to view available options
Quantity:
0.1 g
0.5 g
Unit Size:
0.1g
0.5g
Specifications
Specifications
| Chemical Name or Material | Acenaphthylene D8 |
| CAS | 93951-97-4 |
| Molecular Formula | C12 2H8 |
| Quantity | 0.1 g |
| Synonym | Acenaphthylene-1,2,3,4,5,6,7,8-d8, Acenaphthylene-d8 (9 CI), Acenaphthylene D8 |
| InChI Formula | InChI=1 S/C12H8/c1-3-9-4-2-6-11-8-7-10(5-1)12(9)11/h1-8 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D |
| SMILES | [2 H]C1=C([2 H])c2c([2 H])c([2 H])c([2 H])c3c([2 H])c([2 H])c([2 H])c1c23 |
| IUPAC Name | 1,2,3,4,5,6,7,8-octadeuterioacenaphthylene |
| Molecular Weight (g/mol) | 160.24 |
| Formula Weight | 160.1128 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction