missing translation for 'onlineSavingsMsg'
Learn More

Acenocoumarol, MedChemExpress

Product Code. 16428648
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16428648 10 mg 10mg
16411098 5 mg 5mg
16404527 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16428648 Supplier Medchem Express Supplier No. HYB1014
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Acenocoumarol is an anticoagulant that functions as a Vitamin K antagonist.

Acenocoumarol is an anticoagulant that functions as a Vitamin K antagonist.

Specifications

Chemical Name or Material Acenocoumarol
CAS 152-72-7
Color White
Purity Grade Notes Research
Molecular Formula C19H15NO6
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (283.02 mM; Need ultrasonic)
SMILES O=C1C(C(C2=CC=C([N+]([O-])=O)C=C2)CC(C)=O)=C(O)C3=CC=CC=C3O1
Molecular Weight (g/mol) 353.33
Formula Weight 353.33
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.