missing translation for 'onlineSavingsMsg'
Learn More

Acesulfame Potassium, TRC

Product Code. 17989584
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
100mg
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17989584 1 g 1g
30006522 10 g 10g
17967284 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17989584 Supplier Toronto Research Chemicals Supplier No. TRCA1325001G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55589-62-3
Molecular Formula C4 H4 N O4 S . K
Molecular Weight (g/mol) 201.24
Synonym 1,2,3-Oxathiazin-4(3H)-one, 6-methyl-, 2,2-dioxide, potassium salt (9CI), 6-Methyl-3,4-dihydro-1,2,3-oxathiazin-4-one 2,2-dioxide potassium salt, Acesulfame K, Acesulfame potassium, E 950, Otizon, Potassium 3,4-dihydro-6-methyl-1,2,3-oxathiazine-4-one-2,2-dioxide, Potassium acesulfamate, Potassium acesulfame, Sunett, Sunett D, Sunnett, Sweet One
IUPAC Name potassium;6-methyl-2,2-dioxo-1-oxa-2λ^{6}-thia-3-azanidacyclohex-5-en-4-one
SMILES [K+].CC1=CC(=O)[N-]S(=O)(=O)O1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C4H5NO4S.K/c1-3-2-4(6)5-10(7,8)9-3;/h2H,1H3,(H,5,6);/q;+1/p-1
Formula Weight 200.9498
Percent Purity >95%
Chemical Name or Material Acesulfame potassium
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.