missing translation for 'onlineSavingsMsg'
Learn More

Acetaldehyde dimethyl acetal, 98+%

Product Code. 11404353
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11404353

Brand: Thermo Scientific Alfa Aesar A10939.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 534-15-6 | C4H10O2 | 90.122 g/mol

Acetaldehyde dimethyl acetal may be used in the preparation of glucoside derivatives of steganol. It may be used as polymer solvent for the encapsulation of water-soluble model protein, bovine serum albumin into biodegradable poly(D,L-lactic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetaldehyde dimethyl acetal may be used in the preparation of glucoside derivatives of steganol. It may be used as polymer solvent for the encapsulation of water-soluble model protein, bovine serum albumin into biodegradable poly(D,L-lactic acid.

Solubility
Soluble in water (1000 mg/ml at 25°C), alcohols, chloroform, ether, and acetone.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 534-15-6
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.122
MDL Number MFCD00008493
InChI Key SPEUIVXLLWOEMJ-UHFFFAOYSA-N
Synonym dimethyl acetal, acetaldehyde dimethyl acetal, ethane, 1,1-dimethoxy, dimethylacetal, dimethyl aldehyde, ethylidene dimethyl ether, methyl formyl, acetaldehyde, dimethyl acetal, acetaldehyde methyl acetal, 3-methyl-2,4-dioxapentane
PubChem CID 10795
IUPAC Name 1,1-dimethoxyethane
SMILES CC(OC)OC

Specifications

Melting Point -113°C
Density 0.852
Boiling Point 63°C to 65°C
Flash Point −17°C (1°F)
Odor Sweet
Refractive Index 1.367
Quantity 100 g
UN Number UN2377
Beilstein 1697039
Sensitivity Moisture sensitive
Merck Index 14,3226
Solubility Information Soluble in water (1000mg/ml at 25°C),alcohols,chloroform,ether,and acetone.
Formula Weight 90.12
Percent Purity ≥98%
Chemical Name or Material Acetaldehyde dimethyl acetal
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.