missing translation for 'onlineSavingsMsg'
Learn More

Acetaldehyde ethyl propargyl acetal, 98%, Thermo Scientific™

Product Code. 10025493 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10025493 1 g Glass bottle 1g
10732084 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10025493 Supplier Thermo Scientific Acros Supplier No. 390640010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 18669-04-0
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.17
MDL Number MFCD00066344
InChI Key QKBBTQJLUGADEG-UHFFFAOYSA-N
PubChem CID 86774
IUPAC Name 3-(1-ethoxyethoxy)prop-1-yne
SMILES CCOC(C)OCC#C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8930g/mL
Boiling Point 141°C to 142°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4150 to 1.417
Quantity 1 g
Specific Gravity 0.893
Synonym 3-1-ethoxyethoxy prop-1-yne, acetaldehyde ethyl propargyl acetal, 1-propyne, 3-1-ethoxyethoxy, 1-ethoxy-1-prop-2-yn-1-yloxy ethane, 3-1-ethoxyethoxy propyne, 1-ethoxyethoxy-2-propyne, 1-1-ethoxyethoxy-2propyne, 1-1-ethoxyethoxy-2-propyne, 5-methyl-4,6-dioxa-1-octyne, 1-propyne,3-1-ethoxyethoxy
Formula Weight 128.17
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Acetaldehyde ethyl propargyl acetal
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetaldehyde ethyl propargyl acetal
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.