missing translation for 'onlineSavingsMsg'
Learn More

Acetaldehyde, Extra Pure, SLR, Fisher Chemical™

Product Code. 10152180 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Unit Size:
2.5 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10152180 500 mL 500mL
10497240 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10152180 Supplier Fisher Chemical Supplier No. A/0080/08
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Acetaldehyde, Extra Pure, SLR

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 75-07-0
Molecular Formula C2H4O
MDL Number 6991

Specifikationer

Melting Point -123°C
Boiling Point 21°C
Packaging Amber glass bottle
Quantity 500 mL
Formula Weight 44.04g/mol
Physical Form Liquid
compliance-icons compliance-icons compliance-icons
Produktbeteckning
  • Acetaldehyde
Signalord
  • Danger
Riskkategorier
  • Flammable liquid Category 1
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Carcinogenicity Category 2
Faroangivelser
  • H224-Extremely flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H351-Suspected of causing cancer.
Skyddsangivelse
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P403-Store in a well-ventilated place.
  • P235-Keep cool.
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.