missing translation for 'onlineSavingsMsg'
Learn More

Acetanilide, Extra Pure, SLR, Fisher Chemical™

Product Code. 10407250 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 g
500 g
Unit Size:
250g
500g
Product Code. Quantity unitSize
10407250 250 g 250g
11453473 500 g 500g
2 options
This item is not returnable. View return policy

Product Code. 10407250

Brand: Fisher Chemical A/0200/50

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 103-84-4
MDL Number 8674

Specifications

Melting Point 111°C
Color White
pH 5.0 to 7.0
Boiling Point 304°C
Packaging Plastic powder jar
Molecular Formula C8H9NO
Quantity 250 g
Formula Weight 135.17g/mol
Physical Form Solid
compliance-icons
Product Identifier
  • Acetanilide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.