missing translation for 'onlineSavingsMsg'
Learn More

Acetanilide, Extra Pure, SLR, Fisher Chemical™

Product Code. 11453473 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 g
500 g
Unit Size:
250g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11453473 500 g 500g
10407250 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11453473 Supplier Fisher Chemical Supplier No. A/0200/53
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 103-84-4
Molecular Weight (g/mol) 135.17
MDL Number 8674
InChI Key FZERHIULMFGESH-UHFFFAOYSA-N
IUPAC Name N-phenylacetamide
SMILES CC(=O)NC1=CC=CC=C1

Specifications

Melting Point 111°C
Color White
pH 5.0 to 7.0
Boiling Point 304°C
Packaging Plastic powder jar
Molecular Formula C8H9NO
Quantity 500 g
Formula Weight 135.17g/mol
Physical Form Solid
compliance-icons
Product Identifier
  • Acetanilide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.