missing translation for 'onlineSavingsMsg'
Learn More

Acetamidine hydrochloride, 98+%

Product Code. 10645612 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
10g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10645612 10 g Glass bottle 10g
10438130 100 g Glass bottle 100g
10137803 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10645612 Supplier Thermo Scientific Acros Supplier No. 170210100
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 124-42-5 | C2H6N2·ClH | 94.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-42-5
Molecular Formula C2H6N2·ClH
Molecular Weight (g/mol) 94.54
MDL Number MFCD00013016
InChI Key WCQOBLXWLRDEQA-UHFFFAOYSA-N
Synonym acetamidine hydrochloride, acetimidamide hydrochloride, ethanimidamide hydrochloride, ethanimidamide, monohydrochloride, acetamidine hcl, ethanamidine hydrochloride, acetamidinium chloride, acediamine hydrochloride, ethenylamidine hydrochloride, acetamidine monohydrochloride
PubChem CID 67170
IUPAC Name ethanimidamide;hydrochloride
SMILES CC(=N)N.Cl

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 160°C to 170°C
Color White to Cream
Infrared Spectrum Authentic
Assay Percent Range 98% min. (Argentometry)
Packaging Glass bottle
Linear Formula CH3C(=NH)NH2·HCl
Quantity 10 g
Beilstein 02, 185
Merck Index 15, 45
Solubility Information Solubility in water: 1g/mL. Other solubilities: soluble in alcohol,insoluble in acetone
Formula Weight 94.54
Percent Purity 98+%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material Acetamidine hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.