missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetamiprid-d3, TRC
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Specifications
Specifications
| Chemical Name or Material | Acetamiprid D3 (N-methyl D3) |
| CAS | 1353869-35-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C10 2H3 H8 Cl N4 |
| Quantity | 1 mg |
| Synonym | N-[(6-Chloropyridin-3-yl)methyl]-N'-cyano-N-(trideuteriomethyl)ethanimidamide |
| InChI Formula | InChI=1S/C10H11ClN4/c1-8(14-7-12)15(2)6-9-3-4-10(11)13-5-9/h3-5H,6H2,1-2H3/i2D3 |
| SMILES | [2H]C([2H])([2H])N(Cc1ccc(Cl)nc1)C(=NC#N)C |
| IUPAC Name | N-[(6-chloropyridin-3-yl)methyl]-N'-cyano-N-(trideuteriomethyl)ethanimidamide |
| Molecular Weight (g/mol) | 225.69 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction