missing translation for 'onlineSavingsMsg'
Learn More

Acetanilide, 98%

Product Code. 11456937
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11456937 2500 g 2500g
11374619 100 g 100g
11446937 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11456937 Supplier Thermo Scientific Alfa Aesar Supplier No. A14361.0E
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 103-84-4 | C8H9NO | 135.17 g/mol

Acetanilide is used as an intermediate in the synthesis of rubber accelerator, dyes and camphor. It is also used in the synthesis of penicillin and other pharmaceutical products. It is involved in the preparation of 4-acetamidobenzenesulfonyl chloride, which is an intermediate during the synthesis of sulfa drugs. Further, it is employed as a experimental photographic developer. In addition to this, it is used to stabilize cellulose ester varnishes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetanilide is used as an intermediate in the synthesis of rubber accelerator, dyes and camphor. It is also used in the synthesis of penicillin and other pharmaceutical products. It is involved in the preparation of 4-acetamidobenzenesulfonyl chloride, which is an intermediate during the synthesis of sulfa drugs. Further, it is employed as a experimental photographic developer. In addition to this, it is used to stabilize cellulose ester varnishes.

Solubility
Soluble in ethanol, diethyl ether, acetone, benzene. Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-84-4
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.17
MDL Number MFCD00008674
InChI Key FZERHIULMFGESH-UHFFFAOYSA-N
Synonym acetanilide, acetanil, acetamidobenzene, antifebrin, acetylaniline, acetanilid, n-acetylaniline, acetamide, n-phenyl, acetic acid anilide, acetylaminobenzene
PubChem CID 904
ChEBI CHEBI:28884
IUPAC Name N-phenylacetamide
SMILES CC(=O)NC1=CC=CC=C1

Specifications

Melting Point 113°C to 116°C
Density 1.121
Boiling Point 304°C to 305°C
Flash Point 161°C (321°F)
Assay Percent Range 98%
Quantity 2500 g
Beilstein 606468
Merck Index 14,50
Solubility Information Soluble in ethanol,diethyl ether,acetone,benzene. Slightly soluble in water.
Formula Weight 135.17
Percent Purity 98%
Chemical Name or Material Acetanilide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.