missing translation for 'onlineSavingsMsg'
Learn More

Acetanilide, 99+%

Product Code. 10123440 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
250 g
500 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic bottle
Plastic drum
Unit Size:
1kg
250g
500g
5g
5kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10123440 5 g Glass bottle 5g
10103490 250 g Plastic bottle 250g
10091670 500 g Plastic bottle 500g
10766381 1 kg Plastic bottle 1kg
10445755 5 kg Plastic drum 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10123440 Supplier Thermo Scientific Acros Supplier No. 150810050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 103-84-4 | C8H9NO | 135.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-84-4
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.17
MDL Number MFCD00008674
InChI Key FZERHIULMFGESH-UHFFFAOYSA-N
Synonym acetanilide, acetanil, acetamidobenzene, antifebrin, acetylaniline, acetanilid, n-acetylaniline, acetamide, n-phenyl, acetic acid anilide, acetylaminobenzene
PubChem CID 904
ChEBI CHEBI:28884
IUPAC Name N-phenylacetamide
SMILES CC(=O)NC1=CC=CC=C1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 111.0°C to 115.0°C
Color Blue-Gray to White
Density 4.65 (Air = 1.0)
Boiling Point 304.0°C
Flash Point 173°C
Melting Range ≥111°C to ≤115°C
Infrared Spectrum Conforms to Reference
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3CONHC6H5
Quantity 5 g
Beilstein 12, 237
Merck Index 15, 49
Solubility Information Solubility in water: slightly soluble (5g/L at 25°C). Other solubilities: 1g/20mL boiling water,1g/3.4mL alcohol,1g/4mL ac,etone,1g/3.7mL chloroform,1g/5mL glycerol,1g/8mL,dioxane,1g/18mL diethyl ether,1g/47mL benzene,,sparingly soluble in pe
Formula Weight 135.17
Vapor Pressure 13hPa at 114°C
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material Acetanilide
Show More Show Less
compliance-icons
Product Identifier
  • Acetanilide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.