missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Acetate Buffer (1 M, pH 4.0)

Product Code. 15442939
Change view
Click to view available options
Quantity:
250mL
500mL
Unit Size:
250mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15442939 250mL 250mL
15452939 500mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15442939 Supplier Thermo Scientific Chemicals Supplier No. J60104.AK
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol

Sodium Acetate is used as a buffer in the buffering range of pH 3.6-5.6. Other reported uses. Used in the purification and precipitation of nucleic acids; Used in protein crystallization. Staining of gels in protein gel electrophoresis and HPLC; Mordant dyeing. DNA microarray studies of E. coli response; Investigation of protein unfolding during reverse phase chromatography. Used in conjunction with sodium carbonate to enhance the activation of freeze-dried subtilisin Carlsberg in organic solvents and as a substrate of acetokinase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium Acetate is used as a buffer in the buffering range of pH 3.6-5.6. Other reported uses. Used in the purification and precipitation of nucleic acids; Used in protein crystallization. Staining of gels in protein gel electrophoresis and HPLC; Mordant dyeing. DNA microarray studies of E. coli response; Investigation of protein unfolding during reverse phase chromatography. Used in conjunction with sodium carbonate to enhance the activation of freeze-dried subtilisin Carlsberg in organic solvents and as a substrate of acetokinase.

Solubility
Fully miscible with water. Soluble in ethanol.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-09-3

Specifications

Concentration 1M buffer soln.
pH Range 4
Form Liquid
Quantity 250mL
Storage Requirements Ambient temperatures

RUO – Research Use Only

Product Title
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.