missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Acetate Buffer (1 M, pH 5.5)

Product Code. 15453699
Change view
Click to view available options
Quantity:
250mL
500mL
Unit Size:
250mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15453699 500mL 500mL
15443699 250mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15453699 Supplier Thermo Scientific Chemicals Supplier No. J61033.AP
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol

Acetate, 1M buffer soln., pH, 5.5 it is been widely used in for precipitation of nucleic acids and preparation of gel stains for protein gel electrophoresis. Sodium acetate buffer solution is suitable for precipitation of DNA, elution of proteins and for use in molecular biology applications such as ChIP-on-ChIP assay and in vitro enzyme inhibition assay.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetate, 1M buffer soln., pH, 5.5 it is been widely used in for precipitation of nucleic acids and preparation of gel stains for protein gel electrophoresis. Sodium acetate buffer solution is suitable for precipitation of DNA, elution of proteins and for use in molecular biology applications such as ChIP-on-ChIP assay and in vitro enzyme inhibition assay.

Solubility
Soluble in water (246 g/L) at 20°C, and ethanol.

Notes
Contains sodium acetate and glacial acetic acid, pH 5.5
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-09-3

Specifications

Concentration 1M buffer soln.
Form Liquid
Quantity 500mL
Storage Requirements Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.