missing translation for 'onlineSavingsMsg'
Learn More

Acetate Buffer Solution pH: 5.0, Fisher Chemical™
BIOPHARMA

Product Code. 12960684 Shop All Fisher Chemical Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1000mL
missing translation for 'unitSize'
1000mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12960684 1000mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 12960684 missing translation for 'mfr' Fisher Chemical missing translation for 'supplierNo' SOL 042 1L

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Specifications

pH Range 5.0
Form Liquid
Quantity 1000mL
Product Type Buffer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.