missing translation for 'onlineSavingsMsg'
Learn More

Acetate Buffer Solution pH: 6, Fisher Chemical™
BIOPHARMA

Product Code. 12930694 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1000mL
Unit Size:
1000mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12930694 1000mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12930694 Supplier Fisher Chemical Supplier No. SOL 049 1L
  • ONLINE EXCLUSIVE PRICE
    114.64 €
    103.94 € / 1000mL
    Save 10.70 €
    9% Off
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Specifications

pH Range 6
Form Liquid
Quantity 1000mL
Product Type Buffer
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.