missing translation for 'onlineSavingsMsg'
Learn More

Acetazolamide, 99%

Product Code. 11371947
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11371947 5 g 5g
11381947 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 11371947 Lieferant Thermo Scientific Alfa Aesar Lieferanten-Nr. L07562.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 59-66-5 | C4H6N4O3S2 | 222.24 g/mol

Acetazolamide acts as a carbonic anhydrase inhibitor which increases cerebral blood flow. It inhibits water permeability of membranes by interacting with aquaporins. It is used for the medical treatment of glaucoma, epileptic seizure, idiopathic intracranial hypertension, altitude sickness, cystinuria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in NH4OH (50 mg/mL), DMSO, Methanol and slightly soluble in Ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Incompatible materials are oxidizing agents, bases.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 59-66-5
Molecular Formula C4H6N4O3S2
Molecular Weight (g/mol) 222.24
MDL Number MFCD00003105
InChI Key BZKPWHYZMXOIDC-UHFFFAOYSA-N
Synonym acetazolamide, diamox, acetazolamid, diacarb, glaupax, defiltran, nephramide, acetamox, cidamex, diluran
PubChem CID 1986
ChEBI CHEBI:27690
IUPAC Name N-(5-sulfamoyl-1,3,4-thiadiazol-2-yl)acetamide
SMILES CC(=O)NC1=NN=C(S1)S(N)(=O)=O

Spezifikation

Melting Point ∽255°C (decomposition)
Quantity 5 g
Beilstein 212994
Merck Index 14,53
Solubility Information Soluble in NH4OH (50mg/mL),DMSO,Methanol and slightly soluble in Ethanol.
Formula Weight 222.25
Percent Purity 99%
Chemical Name or Material Acetazolamide

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.