missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, 99.8%, for analysis

Product Code. 10492601 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
5 L
20 L
Packaging:
Glass Bottle
Plastic Can
Plastic drum
Unit Size:
1 L
2.5 L
20 L
250mL
5 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10492601 2.5 L Glass Bottle 2.5 L
10773221 250 mL Glass Bottle 250mL
10492411 1 L Glass Bottle 1 L
10139689 5 L Plastic drum 5 L
10542032 20 L Plastic Can 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10492601 Supplier Thermo Scientific Acros Supplier No. 222140025
  • ONLINE EXCLUSIVE PRICE
    122.00 €
    118.00 € / 2.5 L
    Save 4.00 €
    3% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 64-19-7 | C2H4O2 | 60.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Melting Point 16.0°C to 16.5°C
Boiling Point 117.0°C to 118.0°C
CAS Min % 99.75
CAS Max % 100.0
Density 1.0480g/mL
Quantity 2.5 L
Flash Point 40°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3700 to 1.3730
Residue after Evaporation 0.001% max.
Assay Percent Range 99.8%
Specific Gravity 1.048
Linear Formula CH3COOH
UN Number 2789
Beilstein 02, 96
Fieser 02,5; 05,3; 07,1; 08,1; 12,568; 13,45; 15,12
Merck Index 15, 54
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol, glycerol, ether and, carbon tetrachloride, practically insoluble in carbon disulfide
Formula Weight 60.05
Percent Purity 99.8%
Grade Analytical
Chemical Name or Material Acetic Acid, For Analysis
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetic acid
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.