missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, ammonium salt, 97+%, ACS reagent

Product Code. 10351691 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 kg
Packaging:
Plastic bottle
Unit Size:
25g
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10351691 25 g Plastic bottle 25g
10712222 500 g Plastic bottle 500g
10494701 2 kg Plastic bottle 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10351691 Supplier Thermo Scientific Acros Supplier No. 423270250
  • ONLINE EXCLUSIVE PRICE
    40.10 €
    38.90 € / 25g
    Save 1.20 €
    2% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 631-61-8 | C2H7NO2 | 77.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 631-61-8
Molecular Formula C2H7NO2
Molecular Weight (g/mol) 77.08
MDL Number MFCD00013066
InChI Key USFZMSVCRYTOJT-UHFFFAOYSA-N
Synonym ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
PubChem CID 517165
ChEBI CHEBI:62947
IUPAC Name azanium;acetate
SMILES CC(=O)[O-].[NH4+]

Specifications

Melting Point 114.0°C
Color White
CAS Min % 97.0
CAS Max % 100.0
pH 6.7 to 7.3 (5% soln. at 25°C)
Physical Form Adhering Crystals
Quantity 25 g
Assay Percent Range 97+%
Linear Formula CH3CO2NH4
Beilstein 02, 96
Fieser 01,38; 07,11
Flash Point 136°C
Merck Index 15, 492
Solubility Information Solubility in water: 1480g/L (20°C). Other solubilities: soluble in alcohol, slightly soluble in acetone, in methanol: 7.89 g/100mL (15°C)
Formula Weight 77.08
Percent Purity ≥97%
Grade ACS Reagent
Infrared Spectrum Authentic
Packaging Plastic bottle
Residue after Ignition 0.01% max
Density 1.07
Chemical Name or Material Acetic acid, ammonium salt
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.