missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Acetic acid, ammonium salt, 98%, for biochemistry

Product Code. 12397183 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
12397183 100 g Plastic bottle 100g
12307193 500 g Plastic bottle 500g
12387183 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12397183 Supplier Thermo Scientific Chemicals Supplier No. 447121000
  • ONLINE EXCLUSIVE PRICE
    34.00 €
    31.90 € / 100g
    Save 2.10 €
    6% Off
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 631-61-8 | C2H7NO2 | 77.083 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For biochemistry
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 631-61-8
Molecular Formula C2H7NO2
Molecular Weight (g/mol) 77.083
InChI Key USFZMSVCRYTOJT-UHFFFAOYSA-N
Synonym ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
PubChem CID 517165
ChEBI CHEBI:62947
IUPAC Name azanium;acetate
SMILES CC(=O)[O-].[NH4+]

Specifications

Absorbance (c=1M, water), (c=1M, water) at 0nm, 0.01 max. at 280nm, 0.01 max. at 350nm, 0.02 max. at 254nm
Melting Point 114.0°C
Quantity 100 g
Flash Point 136°C
Packaging Plastic bottle
Assay Percent Range 98%
Linear Formula CH3CO2NH4
Solubility Information Solubility in water: 1480g/L (20°C). Other solubilities: soluble in alcohol, slightly soluble in acetone, in methanol: 7.89 g/100mL (15°C)
Formula Weight 77.08
Percent Purity 98%
Grade Biochemical
Chemical Name or Material Acetic acid, ammonium salt
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.