missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid-d4 (Reagent grade), CDN

Product Code. 30320533
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30320533 10 g 10g
30317583 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30320533 Supplier CDN Supplier No. CDND000110G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acetic acid-D4
CAS 1186-52-3
Molecular Formula C2 2H4 O2
Quantity 10 g
InChI Formula InChI=1 S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)/i1D3/hD
SMILES [2 H]OC(=O)C([2 H])([2 H])[2 H]
IUPAC Name deuterio 2,2,2-trideuterioacetate
Molecular Weight (g/mol) 64.08
Formula Weight 64.0462 g/mol
Percent Purity 99 atom % D, min 99% Chemical Purity
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.