missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid-d4, TRC

Product Code. 30327348
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30327348 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30327348 Supplier Toronto Research Chemicals Supplier No. TRCA16765725G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acetic acid-D4
CAS 1186-52-3
Molecular Formula C2 2H4 O2
Quantity 25 g
Synonym Acetic-d3 acid-d (6CI,7CI,8CI,9CI), Acetic acid (CD3COOD), Acetic acid-d4, Perdeuterioacetic acid, Perdeuteroacetic acid, Tetradeuterioacetic acid, Tetradeuteroacetic acid, Acetic acid D4
InChI Formula InChI=1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)/i1D3/hD
SMILES [2H]OC(=O)C([2H])([2H])[2H]
IUPAC Name deuterio 2,2,2-trideuterioacetate
Molecular Weight (g/mol) 64.08
Formula Weight 64.0462
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.