missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic acid glacial 99-100%, BAKER ANALYZED™ ACS, J.T. Baker™
Shop All J.T.Baker Products
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Plastic bottle
Unit Size:
1 L
2.5 L
Specifications
Specifications
| Color | Max. 10 (APHA) |
| Assay Percent Range | min. 99.7% |
| Quantity | 1 L |
| Packaging | Plastic bottle |
| Residue after Evaporation | max. 0.001% |
Safety and Handling
Signal Word
- Danger
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 A
- H226-Flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction