missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, Glacial (Aldehyde-Free/Sequencing), Fisher BioReagents™™

Product Code. 10021123
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Glass
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10021123 500 mL Glass 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10021123 Supplier Fisher BioReagents Supplier No. BP1185500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For DNA sequencing and other biological applications such as DNA/RNA electrophoresis | CAS: 64-19-7 | C2H4O2 | 60.05 g/mol

For DNA sequencing and other biological applications such as DNA/RNA electrophoresis

Aldehyde-free Acetic Acid is suitable for DNA sequencing and other biological applications.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acetic Acid, Glacial
Name Note Aldehyde-free
pH 2.5
CAS 64-19-7
Melting Point 16°C
Boiling Point 117°C
Color Colorless
Assay Percent Range ≥99.7 %
Molecular Formula C2H4O2
Linear Formula CH3COOH
MDL Number MFCD00036152
UN Number 2789
Merck Index 15, 54
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
SMILES CC(O)=O
IUPAC Name acetic acid
Molecular Weight (g/mol) 60.05
PubChem CID 176
ChEBI CHEBI:15366
Formula Weight 60.05
Percent Purity ≥99.7%
Grade Sequencing
Density 1.048g/cm³
Quantity 500 mL
Packaging Glass
Physical Form Liquid
Residue after Evaporation 8ppm max.
Vapor Pressure 1.52 kPa at 20°C
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetic acid
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.