missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, Glacial, for HPLC, Thermo Scientific
GREENER_CHOICE

Product Code. 30280928 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30280928 1 L 1 L
30280927 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30280928 Supplier Thermo Scientific Supplier No. T004061000
  • ONLINE EXCLUSIVE PRICE
    106.00 €
    78.00 € / 1 L
    Save 28.00 €
    26% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

Molecular Weight (g/mol) 60.05 g/mol

Specifications

CAS 64-19-7
Grade HPLC
Quantity 1 L
Packaging Plastic Coated Glass Bottle
Chemical Name or Material Acetic Acid, Glacial
compliance-icons compliance-icons
Product Identifier
  • Acetic acid, glacial
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Acetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.