missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, potassium salt, 99+%, for molecular biology

Product Code. 10254644 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
100g
1kg
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10254644 250 g Glass bottle 250g
10163114 100 g Glass bottle 100g
10103074 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10254644 Supplier Thermo Scientific Acros Supplier No. 397122500
  • ONLINE EXCLUSIVE PRICE
    58.65 €
    55.00 € / 250g
    Save 3.65 €
    6% Off
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 127-08-2 | C2H3KO2 | 98.142 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer for molecular biology
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-08-2
Molecular Formula C2H3KO2
Molecular Weight (g/mol) 98.142
MDL Number MFCD00012458
InChI Key SCVFZCLFOSHCOH-UHFFFAOYSA-M
Synonym potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
PubChem CID 517044
ChEBI CHEBI:32029
IUPAC Name potassium;acetate
SMILES CC(=O)[O-].[K+]

Specifications

Melting Point 292.0°C
CAS Min % 99.0
CAS Max % 100.0
Color White to Brown
Quantity 250 g
Flash Point >250°C
Packaging Glass bottle
Physical Form Low Melting Solid
Assay Percent Range 99+%
Linear Formula CH3CO2K
Merck Index 15, 7726
Solubility Information Solubility in water: 2694g/L (25°C). Other solubilities: soluble in alcohol, insoluble in ether
Formula Weight 98.14
Percent Purity ≥99%
Grade Molecular Biology
Chemical Name or Material Acetic acid, potassium salt
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.