missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.8%, Solstice

Product Code. 15633920
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
30 kg
4 x 2.5 L
60 kg
6 x 1 L
200 L
Packaging:
Glass Bottle
Plastic Bottle
Plastic Pail
Poly Drum
Unit Size:
1 L
2.5 L
200 L
30kg
5 L
60kg
Pack of 4
Pack of 6
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15633920 1 L Plastic Bottle 1 L
15643920 1 L Glass Bottle 1 L
15663920 2.5 L Glass Bottle 2.5 L
15653920 2.5 L Plastic Bottle 2.5 L
15613930 5 L Plastic Bottle 5 L
15643930 6 x 1 L Glass Bottle Pack of 6
15633930 6 x 1 L Plastic Bottle Pack of 6
15603930 4 x 2.5 L Glass Bottle Pack of 4
15693920 4 x 2.5 L Plastic Bottle Pack of 4
15683920 30 kg Plastic Pail 30kg
15623930 60 kg Plastic Pail 60kg
15673920 200 L Poly Drum 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Product Code. 15633920 Supplier Honeywell-Fluka Supplier No. 332091L
  • ONLINE EXCLUSIVE PRICE
    69.00 €
    12.29 € / 1 L
    Save 56.71 €
    82% Off
View all promotions
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.8%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Density 1.049g/mL at 25°C
Quantity 1 L
Explosion Limit 16% at 92°F, 4% at 59°F
Flash Point 40°C (104°F)
Ignition Point 800°F
Packaging Plastic Bottle
Linear Formula CH3CO2H
Vapor Density 2.07 (vs air)
UN Number UN2789
Beilstein 506007
Formula Weight 60.05g/mol
Percent Purity ≥99.8%
Grade ACS/puriss. p.a.
Chemical Name or Material Acetic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetic acid
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
Supplemental information
  • MIXTURE LIST-Contain: acetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.